Assay Details
| Gene Symbol: dgkaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045315.1 | NP_001038780.1 | 7 - 8 | 684 | 62 | |
| XM_005162328.4 | XP_005162385.1 | 8 - 9 | 838 | 62 | ||
| GenBank mRNA | BC098607.1 | - | 7 - 8 | 680 | 62 | |
| BC117645.1 | - | 7 - 8 | 684 | 62 | ||
More Information
Set Membership: |
|

dgkaa
Genomic Map