Assay Details
| Gene Symbol: dgkaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045315.1 | NP_001038780.1 | 9 - 10 | 915 | 57 | |
| XM_005162328.4 | XP_005162385.1 | 10 - 11 | 1069 | 57 | ||
| GenBank mRNA | BC098607.1 | - | 9 - 10 | 911 | 57 | |
| BC117645.1 | - | 9 - 10 | 915 | 57 | ||
| FJ392635.1 | - | 1 - 2 | 60 | 57 | ||
More Information
Set Membership: |
|

dgkaa
Genomic Map