Assay Details
| Gene Symbol: gfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077746.2 | NP_001071214.2 | 2 - 3 | 645 | 64 | |
| GenBank mRNA | BC125879.1 | - | 2 - 3 | 582 | 64 | |
More Information
Set Membership: |
|

gfod2
Genomic Map