Assay Details
| Gene Symbol: prmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080175.2 | NP_001073644.2 | 3 - 4 | 547 | 69 | |
| XM_021478690.1 | XP_021334365.1 | 2 - 3 | 374 | 69 | ||
| GenBank mRNA | BC129171.1 | - | 3 - 4 | 488 | 69 | |
| CT641169.2 | - | 3 - 4 | 516 | 69 | ||
More Information
Set Membership: |
|

prmt2
Genomic Map