Assay Details
| Gene Symbol: ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313685.1 | NP_001300614.1 | 3 - 4 | 776 | 77 | |
| NM_001313686.1 | NP_001300615.1 | 3 - 4 | 813 | 77 | ||
| NM_130924.3 | NP_570999.2 | 3 - 4 | 863 | 77 | ||
| GenBank mRNA | BC049294.1 | - | 3 - 4 | 776 | 77 | |
| BC054648.1 | - | 756 | 77 | |||
| BC164734.1 | - | 724 | 77 | |||
| FN428729.1 | - | 3 - 4 | 368 | 77 | ||
| Y14427.1 | - | 863 | 77 | |||
More Information
Set Membership: |
|

ptpn1
Genomic Map