Assay Details
| Gene Symbol: dbx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131178.1 | NP_571253.1 | 1 - 2 | 439 | 109 | |
| GenBank mRNA | AF030285.1 | - | 1 - 2 | 439 | 109 | |
| BC162275.1 | - | 1 - 2 | 439 | 109 | ||
| BC162291.1 | - | 1 - 2 | 439 | 109 | ||
More Information
Set Membership: |
|

dbx1b
Genomic Map