Assay Details
| Gene Symbol: foxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131271.2 | NP_571346.2 | 1 | 1468 | 73 | |
| GenBank mRNA | AF052252.1 | - | 1 | 1025 | 73 | |
| BC075922.1 | - | 1 | 1451 | 73 | ||
More Information
Set Membership: |
|

foxd2
Genomic Map