Assay Details
| Gene Symbol: atp1b2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131838.2 | NP_571913.1 | 3 - 4 | 635 | 108 | |
| XM_005165260.3 | XP_005165317.1 | 3 - 4 | 654 | 108 | ||
| GenBank mRNA | AF373976.1 | - | 3 - 4 | 642 | 108 | |
| BC064702.1 | - | 3 - 4 | 635 | 108 | ||
More Information
Set Membership: |
|

atp1b2b
Genomic Map