Assay Details
| Gene Symbol: atp1b2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131838.2 | NP_571913.1 | 5 - 6 | 905 | 103 | |
| GenBank mRNA | AF373976.1 | - | 5 - 6 | 912 | 103 | |
| BC064702.1 | - | 5 - 6 | 905 | 103 | ||
More Information
Set Membership: |
|

atp1b2b
Genomic Map