Assay Details
| Gene Symbol: prickle1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183342.2 | NP_899185.2 | 2 - 3 | 330 | 84 | |
| XM_005174210.4 | XP_005174267.1 | 2 - 3 | 419 | 84 | ||
| GenBank mRNA | AY278986.1 | - | 1 - 2 | 149 | 84 | |
| AY286492.1 | - | 2 - 3 | 330 | 84 | ||
| BC162516.1 | - | 1 - 2 | 176 | 84 | ||
More Information
Set Membership: |
|

prickle1a
Genomic Map