Assay Details
| Gene Symbol: prickle1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183342.2 | NP_899185.2 | 3 - 4 | 442 | 97 | |
| XM_005174210.4 | XP_005174267.1 | 3 - 4 | 531 | 97 | ||
| GenBank mRNA | AY278986.1 | - | 2 - 3 | 261 | 97 | |
| AY286492.1 | - | 3 - 4 | 442 | 97 | ||
| BC162516.1 | - | 2 - 3 | 288 | 97 | ||
More Information
Set Membership: |
|

prickle1a
Genomic Map