Assay Details
| Gene Symbol: slc52a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199656.1 | NP_955950.1 | 4 - 5 | 1226 | 67 | |
| GenBank mRNA | BC055241.1 | - | 4 - 5 | 1226 | 67 | |
More Information
Set Membership: |
|

slc52a2
Genomic Map