Assay Details
| Gene Symbol: stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200120.1 | NP_956414.1 | 5 - 6 | 597 | 64 | |
| XM_005163455.4 | XP_005163512.1 | 5 - 6 | 698 | 64 | ||
| GenBank mRNA | BC045442.1 | - | 5 - 6 | 597 | 64 | |
| BC164220.1 | - | 5 - 6 | 476 | 64 | ||
More Information
Set Membership: |
|

stam
Genomic Map