Assay Details
| Gene Symbol: vrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201170.2 | NP_957464.2 | 11 - 12 | 1415 | 62 | |
| XM_017358726.2 | XP_017214215.1 | 10 - 11 | 1477 | 62 | ||
| GenBank mRNA | BC049319.1 | - | 11 - 12 | 1366 | 62 | |
More Information
Set Membership: |
|

vrk2
Genomic Map