Assay Details
| Gene Symbol: ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201292.1 | NP_958449.1 | 5 - 6 | 710 | 85 | |
| XM_009307281.3 | XP_009305556.1 | 5 - 6 | 795 | 85 | ||
| XM_021480492.1 | XP_021336167.1 | 5 - 6 | 699 | 85 | ||
| GenBank mRNA | AY394946.1 | - | 5 - 6 | 710 | 85 | |
| BC128809.1 | - | 5 - 6 | 716 | 85 | ||
| BC155777.1 | - | 6 - 7 | 851 | 85 | ||
More Information
Set Membership: |
|

ap3m1
Genomic Map