Assay Details
| Gene Symbol: sec61a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201577.1 | NP_963871.1 | 11 - 12 | 1314 | 60 | |
| GenBank mRNA | AY034614.1 | - | 11 - 12 | 1314 | 60 | |
| BC048881.1 | - | 11 - 12 | 1315 | 60 | ||
| BC165352.1 | - | 10 - 11 | 1280 | 60 | ||
More Information
Set Membership: |
|

sec61a1l
Genomic Map