Assay Details
| Gene Symbol: dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205627.2 | NP_991190.2 | 8 - 9 | 1954 | 56 | |
| XM_005168270.4 | XP_005168327.1 | 9 - 10 | 2436 | 56 | ||
| GenBank mRNA | AY208855.1 | - | 8 - 9 | 1909 | 56 | |
| AY429681.1 | - | 8 - 9 | 1954 | 56 | ||
| AY641096.1 | - | 9 - 10 | 2038 | 56 | ||
| BC067573.1 | - | 6 - 7 | 872 | 56 | ||
| BC118677.1 | - | 9 - 10 | 2424 | 56 | ||
More Information
Set Membership: |
|

dstyk
Genomic Map