Assay Details
| Gene Symbol: gadd45ga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205691.1 | NP_991254.1 | 2 - 3 | 207 | 72 | |
| GenBank mRNA | AY714219.1 | - | 2 - 3 | 186 | 72 | |
| BC065870.1 | - | 2 - 3 | 207 | 72 | ||
More Information
Set Membership: |
|

gadd45ga
Genomic Map