Assay Details
| Gene Symbol: fam49bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207078.1 | NP_996961.1 | 5 - 6 | 457 | 109 | |
| GenBank mRNA | BC066542.1 | - | 5 - 6 | 457 | 109 | |
| BC152197.1 | - | 5 - 6 | 521 | 109 | ||
More Information
Set Membership: |
|

fam49bb
Genomic Map