Assay Details
| Gene Symbol: fam49bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207078.1 | NP_996961.1 | 10 - 11 | 996 | 72 | |
| GenBank mRNA | BC066542.1 | - | 10 - 11 | 996 | 72 | |
| BC152197.1 | - | 10 - 11 | 1060 | 72 | ||
More Information
Set Membership: |
|

fam49bb
Genomic Map