Assay Details
| Gene Symbol: per1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212439.2 | NP_997604.2 | 10 - 11 | 2446 | 77 | |
| GenBank mRNA | AY597250.1 | - | 10 - 11 | 2446 | 77 | |
| BC163718.1 | - | 9 - 10 | 1991 | 77 | ||
More Information
Set Membership: |
|

per1b
Genomic Map