Assay Details
| Gene Symbol: tpd52l2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328541.1 | NP_001315470.1 | 2 - 3 | 281 | 66 | |
| NM_001328542.1 | NP_001315471.1 | 2 - 3 | 281 | 66 | ||
| NM_001328543.1 | NP_001315472.1 | 2 - 3 | 281 | 66 | ||
| NM_212733.2 | NP_997898.1 | 2 - 3 | 281 | 66 | ||
| XM_005167010.4 | XP_005167067.1 | 2 - 3 | 256 | 66 | ||
| XM_009303929.3 | XP_009302204.1 | 2 - 3 | 251 | 66 | ||
| XM_021478189.1 | XP_021333864.1 | 2 - 3 | 251 | 66 | ||
| GenBank mRNA | BC044439.1 | - | 2 - 3 | 281 | 66 | |
| BC067337.1 | - | 2 - 3 | 215 | 66 | ||
| EH435837.1 | - | 2 - 3 | 263 | 66 | ||
More Information
Set Membership: |
|

tpd52l2a
Genomic Map