Assay Details
| Gene Symbol: prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213496.1 | NP_998661.1 | 7 - 8 | 677 | 61 | |
| XM_005164119.3 | XP_005164176.1 | 7 - 8 | 783 | 61 | ||
| XM_005164120.4 | XP_005164177.1 | 6 - 7 | 641 | 61 | ||
| XM_021470043.1 | XP_021325718.1 | 8 - 9 | 945 | 61 | ||
| GenBank mRNA | BC044179.1 | - | 7 - 8 | 680 | 61 | |
| BC067543.1 | - | 7 - 8 | 677 | 61 | ||
More Information
Set Membership: |
|

prpsap2
Genomic Map