Assay Details
| Gene Symbol: asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214730.1 | NP_999895.1 | 3 - 4 | 367 | 67 | |
| XM_005172174.4 | XP_005172231.1 | 3 - 4 | 412 | 67 | ||
| XM_005172175.4 | XP_005172232.1 | 3 - 4 | 415 | 67 | ||
| GenBank mRNA | BC065964.1 | - | 3 - 4 | 367 | 67 | |
More Information
Set Membership: |
|

asb7
Genomic Map