Assay Details
| Gene Symbol: cyp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001730.2 | NP_001001730.2 | 2 - 3 | 345 | 118 | |
| GenBank mRNA | BC116552.1 | - | 2 - 3 | 345 | 118 | |
| BC164033.1 | - | 2 - 3 | 288 | 118 | ||
More Information
Set Membership: |
|

cyp51
Genomic Map