Assay Details
| Gene Symbol: rpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002078.3 | NP_001002078.1 | 2 - 3 | 182 | 71 | |
| GenBank mRNA | BC072550.1 | - | 1 - 2 | 108 | 71 | |
| EH452858.1 | - | 1 - 2 | 149 | 71 | ||
| EH544684.1 | - | 2 - 3 | 218 | 71 | ||
| EH544712.1 | - | 2 - 3 | 170 | 71 | ||
More Information
Set Membership: |
|

rpl39
Genomic Map