Assay Details
| Gene Symbol: mettl21e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002361.2 | NP_001002361.2 | 2 - 3 | 213 | 106 | |
| GenBank mRNA | BC076050.1 | - | 1 - 2 | 176 | 106 | |
| EB850753.1 | - | 2 - 3 | 204 | 106 | ||
More Information
Set Membership: |
|

mettl21e
Genomic Map