Assay Details
| Gene Symbol: sult1st6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002599.1 | NP_001002599.1 | 3 - 4 | 719 | 72 | |
| GenBank mRNA | AY937249.1 | - | 3 - 4 | 405 | 72 | |
| BC075996.1 | - | 3 - 4 | 719 | 72 | ||
More Information
Set Membership: |
|

sult1st6
Genomic Map