Assay Details
| Gene Symbol: sult1st6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002599.1 | NP_001002599.1 | 4 - 5 | 857 | 81 | |
| GenBank mRNA | AY937249.1 | - | 4 - 5 | 543 | 81 | |
| BC075996.1 | - | 4 - 5 | 857 | 81 | ||
More Information
Set Membership: |
|

sult1st6
Genomic Map