Assay Details
| Gene Symbol: elac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003503.1 | NP_001003503.1 | 2 - 3 | 702 | 61 | |
| XM_021476083.1 | XP_021331758.1 | 2 - 3 | 888 | 61 | ||
| GenBank mRNA | BC078352.1 | - | 2 - 3 | 702 | 61 | |
More Information
Set Membership: |
|

elac1
Genomic Map