Assay Details
| Gene Symbol: cnot3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328118.1 | NP_001315047.1 | 14 - 15 | 2321 | 80 | |
| XM_005158349.4 | XP_005158406.1 | 14 - 15 | 2325 | 80 | ||
| XM_021466678.1 | XP_021322353.1 | 14 - 15 | 2340 | 80 | ||
| GenBank mRNA | BC081678.1 | - | 11 - 12 | 1371 | 80 | |
More Information
Set Membership: |
|

cnot3a
Genomic Map