Assay Details
| Gene Symbol: scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005776.1 | NP_001005776.1 | 5 - 6 | 454 | 67 | |
| XM_021466744.1 | XP_021322419.1 | 5 - 6 | 484 | 67 | ||
| GenBank mRNA | BC083168.1 | - | 5 - 6 | 454 | 67 | |
More Information
Set Membership: |
|

scgn
Genomic Map