Assay Details
| Gene Symbol: fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007315.1 | NP_001007316.1 | 3 - 4 | 1028 | 64 | |
| GenBank mRNA | BC085657.1 | - | 3 - 4 | 1028 | 64 | |
More Information
Set Membership: |
|

fbxl4
Genomic Map