Assay Details
| Gene Symbol: mcamb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012367.2 | NP_001012367.2 | 3 - 4 | 510 | 86 | |
| XM_005157570.2 | XP_005157627.1 | 3 - 4 | 533 | 86 | ||
| XM_021481353.1 | XP_021337028.1 | 3 - 4 | 533 | 86 | ||
| GenBank mRNA | AY497061.1 | - | 3 - 4 | 489 | 86 | |
| AY746594.1 | - | 3 - 4 | 510 | 86 | ||
| BC162589.1 | - | 3 - 4 | 502 | 86 | ||
More Information
Set Membership: |
|

mcamb
Genomic Map