Assay Details
| Gene Symbol: fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017636.1 | NP_001017636.1 | 2 - 3 | 372 | 69 | |
| GenBank mRNA | BC092815.1 | - | 2 - 3 | 372 | 69 | |
More Information
Set Membership: |
|

fam50a
Genomic Map