Assay Details
| Gene Symbol: tprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030123.1 | NP_001025294.1 | 13 - 14 | 1514 | 101 | |
| XM_009294751.3 | XP_009293026.1 | 13 - 14 | 1535 | 101 | ||
| GenBank mRNA | BC134977.1 | - | 13 - 14 | 1512 | 101 | |
More Information
Set Membership: |
|

tprb
Genomic Map