Assay Details
| Gene Symbol: tprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030123.1 | NP_001025294.1 | 31 - 32 | 4531 | 62 | |
| XM_009294751.3 | XP_009293026.1 | 31 - 32 | 4552 | 62 | ||
More Information
Set Membership: |
|

tprb
Genomic Map