Assay Details
| Gene Symbol: nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037114.2 | NP_001032191.2 | 5 - 6 | 680 | 87 | |
| NM_001312901.1 | NP_001299830.1 | 4 - 5 | 561 | 87 | ||
| GenBank mRNA | BC107506.1 | - | 4 - 5 | 622 | 87 | |
| EH446733.1 | - | 5 - 6 | 644 | 88 | ||
| EH482962.1 | - | 2 - 3 | 388 | 87 | ||
| EH542357.1 | - | 5 - 6 | 750 | 88 | ||
More Information
Set Membership: |
|

nubp2
Genomic Map