Assay Details
| Gene Symbol: sox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037680.1 | NP_001032769.1 | 1 | 1048 | 67 | |
| GenBank mRNA | BC108033.1 | - | 1 | 1048 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sox14| Gene Symbol: sox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037680.1 | NP_001032769.1 | 1 | 1048 | 67 | |
| GenBank mRNA | BC108033.1 | - | 1 | 1048 | 67 | |
Set Membership: |
|