Assay Details
| Gene Symbol: ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037705.1 | NP_001032794.1 | 2 - 3 | 408 | 58 | |
| GenBank mRNA | BC108061.1 | - | 2 - 3 | 408 | 58 | |
More Information
Set Membership: |
|

ccdc92
Genomic Map