Assay Details
| Gene Symbol: col18a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ967004.1 | - | 1508 | 71 | ||
| AJ967005.1 | - | 1610 | 71 | |||
| AJ967006.1 | - | 2396 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

col18a1a| Gene Symbol: col18a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ967004.1 | - | 1508 | 71 | ||
| AJ967005.1 | - | 1610 | 71 | |||
| AJ967006.1 | - | 2396 | 71 | |||
Set Membership: |
|