Assay Details
| Gene Symbol: col18a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001349195.1 | NP_001336124.1 | 9 - 10 | 1397 | 80 | |
| XM_021478720.1 | XP_021334395.1 | 9 - 10 | 2198 | 80 | ||
| XM_021478721.1 | XP_021334396.1 | 8 - 9 | 1819 | 80 | ||
| XM_021478722.1 | XP_021334397.1 | 10 - 11 | 1422 | 80 | ||
| GenBank mRNA | AJ967004.1 | - | 9 - 10 | 1245 | 81 | |
| AJ967005.1 | - | 9 - 10 | 1347 | 81 | ||
| AJ967006.1 | - | 9 - 10 | 2133 | 81 | ||
| BC163779.1 | - | 9 - 10 | 1397 | 80 | ||
More Information
Set Membership: |
|

col18a1a
Genomic Map