Assay Details
| Gene Symbol: acbd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100088.1 | NP_001093558.1 | 8 - 9 | 784 | 74 | |
| XM_005171138.3 | XP_005171195.1 | 8 - 9 | 835 | 74 | ||
| XM_005171140.2 | XP_005171197.1 | 8 - 9 | 814 | 74 | ||
| XM_021478275.1 | XP_021333950.1 | 8 - 9 | 814 | 74 | ||
| XM_021478280.1 | XP_021333955.1 | 7 - 8 | 618 | 74 | ||
More Information
Set Membership: |
|

acbd5b
Genomic Map