Assay Details
| Gene Symbol: map3k22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105691.1 | NP_001099161.1 | 11 - 12 | 983 | 68 | |
| XM_005162667.4 | XP_005162724.1 | 11 - 12 | 1372 | 68 | ||
| GenBank mRNA | BC152572.1 | - | 11 - 12 | 983 | 68 | |
More Information
Set Membership: |
|

map3k22
Genomic Map