Assay Details
| Gene Symbol: map3k22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105691.1 | NP_001099161.1 | 12 - 13 | 1060 | 64 | |
| GenBank mRNA | BC152572.1 | - | 12 - 13 | 1060 | 64 | |
More Information
Set Membership: |
|

map3k22
Genomic Map