Assay Details
| Gene Symbol: calm3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182967.1 | NP_892012.1 | 4 - 5 | 391 | 123 | |
| GenBank mRNA | BC045298.1 | - | 4 - 5 | 391 | 123 | |
| BC071404.1 | - | 4 - 5 | 417 | 123 | ||
| BC164893.1 | - | 3 - 4 | 313 | 123 | ||
More Information
Set Membership: |
|

calm3a
Genomic Map