Assay Details
| Gene Symbol: usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199579.1 | NP_955873.1 | 2 - 3 | 271 | 75 | |
| GenBank mRNA | BC045507.1 | - | 2 - 3 | 271 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

usp1| Gene Symbol: usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199579.1 | NP_955873.1 | 2 - 3 | 271 | 75 | |
| GenBank mRNA | BC045507.1 | - | 2 - 3 | 271 | 75 | |
Set Membership: |
|