Assay Details
| Gene Symbol: cnot3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199840.1 | NP_956134.1 | 12 - 13 | 2033 | 131 | |
| XM_009294029.3 | XP_009292304.1 | 12 - 13 | 2053 | 131 | ||
| XM_017352187.2 | XP_017207676.1 | 12 - 13 | 2046 | 131 | ||
| GenBank mRNA | AY391415.1 | - | 12 - 13 | 1945 | 131 | |
| BC045499.1 | - | 12 - 13 | 2033 | 131 | ||
More Information
Set Membership: |
|

cnot3b
Genomic Map