Assay Details
| Gene Symbol: cnot3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199840.1 | NP_956134.1 | 7 - 8 | 656 | 83 | |
| XM_009294029.3 | XP_009292304.1 | 7 - 8 | 676 | 83 | ||
| XM_017352187.2 | XP_017207676.1 | 7 - 8 | 669 | 83 | ||
| GenBank mRNA | BC045499.1 | - | 7 - 8 | 656 | 83 | |
More Information
Set Membership: |
|

cnot3b
Genomic Map