Assay Details
| Gene Symbol: got2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199989.1 | NP_956283.1 | 5 - 6 | 652 | 83 | |
| GenBank mRNA | BC049435.1 | - | 5 - 6 | 652 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

got2b| Gene Symbol: got2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199989.1 | NP_956283.1 | 5 - 6 | 652 | 83 | |
| GenBank mRNA | BC049435.1 | - | 5 - 6 | 652 | 83 | |
Set Membership: |
|